3-(3-Methoxyphenoxy)azetidine

Supplier: Matrix Scientific

049515-500MG 049515-2.5G
101887-964EA 364.62 USD
101887-964 101887-966
3-(3-Methoxyphenoxy)azetidine
3-(3-Methoxyphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD09756542
CAS Number: 954220-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR