N-[3-(3-Azetidinyloxy)phenyl]acetamide hydrochloride

Supplier: Matrix Scientific

049514-500MG 049514-2.5G
101887-960EA 364.62 USD
101887-960 101887-962
N-[3-(3-Azetidinyloxy)phenyl]acetamide hydrochloride
N-[3-(3-Azetidinyloxy)phenyl]acetamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₂
MDL Number: MFCD28714455
CAS Number: 1820736-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR