3-(3,4-Dichlorophenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049513-500MG 049513-2.5G
101887-956EA 166.15 USD
101887-956 101887-958
3-(3,4-Dichlorophenoxy)azetidine hydrochloride
3-(3,4-Dichlorophenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₀Cl₃NO
MDL Number: MFCD09264481
CAS Number: 606129-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR