3-(3-Chlorophenoxy)azetidine hydrochloride
Supplier: Matrix Scientific
049512-500MG
049512-2.5G
101887-952EA
364.62
USD
101887-952
101887-954
3-(3-Chlorophenoxy)azetidine hydrochloride
3-(3-Chlorophenoxy)azetidine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁Cl₂NO MW: 220.1 g/mol |
MDL Number:
MFCD20233672 CAS Number: 868833-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...