2-Allylphenyl-3-azetidinyl ether

Supplier: Matrix Scientific

049508-500MG 049508-2.5G
101887-936EA 364.62 USD
101887-936 101887-938
2-Allylphenyl-3-azetidinyl ether
2-Allylphenyl-3-azetidinyl ether

Hazard: Irritant

Formula: C₁₂H₁₅NO
MDL Number: MFCD13559944
CAS Number: 1220028-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR