3-(2,4-Dimethylphenoxy)azetidine

Supplier: Matrix Scientific

049504-500MG 049504-2.5G
101887-920EA 364.62 USD
101887-920 101887-922
3-(2,4-Dimethylphenoxy)azetidine
3-(2,4-Dimethylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD09861875
CAS Number: 954223-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR