3-(2-Methylphenoxy)azetidine
Supplier: Matrix Scientific
Synonyms:
3-(2-Methylphenoxy)azetidine hydrochloride
049501-500MG
049501-2.5G
101887-908EA
364.62
USD
101887-908
101887-910
3-(2-Methylphenoxy)azetidine
3-(2-Methylphenoxy)azetidine
Hazard: Irritant
Formula:
C₁₀H₁₃NO MW: 163.22 g/mol |
MDL Number:
MFCD09861857 CAS Number: 954222-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...