3-[2-(sec-Butyl)phenoxy]azetidine

Supplier: Matrix Scientific

049500-500MG 049500-2.5G
101887-904EA 364.62 USD
101887-904 101887-906
3-[2-(sec-Butyl)phenoxy]azetidine
3-[2-(sec-Butyl)phenoxy]azetidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD13559939
CAS Number: 1220028-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR