3-[2-(tert-Butyl)phenoxy]azetidine

Supplier: Matrix Scientific

049498-500MG 049498-2.5G
101887-896EA 364.62 USD
101887-896 101887-898
3-[2-(tert-Butyl)phenoxy]azetidine
3-[2-(tert-Butyl)phenoxy]azetidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD13559937
CAS Number: 1146956-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR