3-[2-(Trifluoromethyl)phenoxy]azetidine hydrochloride

Supplier: Matrix Scientific

049497-500MG 049497-2.5G
101887-892EA 384.62 USD
101887-892 101887-894
3-[2-(Trifluoromethyl)phenoxy]azetidine hydrochloride
3-[2-(Trifluoromethyl)phenoxy]azetidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₁ClF₃NO
MDL Number: MFCD11849028
CAS Number: 82622-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR