3-([1,1'-Biphenyl]-2-yloxy)azetidine

Supplier: Matrix Scientific

049496-500MG 049496-2.5G
101887-888EA 364.62 USD
101887-888 101887-890
3-([1,1'-Biphenyl]-2-yloxy)azetidine
3-([1,1'-Biphenyl]-2-yloxy)azetidine

Hazard: Irritant

Formula: C₁₅H₁₅NO
MDL Number: MFCD13559936
CAS Number: 1219948-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR