3-(2-Methoxyphenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049494-500MG 049494-2.5G
101887-880EA 384.62 USD
101887-880 101887-882
3-(2-Methoxyphenoxy)azetidine hydrochloride
3-(2-Methoxyphenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD13186326
CAS Number: 954223-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR