3-(2-Chloro-5-methylphenoxy)azetidine

Supplier: Matrix Scientific

049493-500MG 049493-2.5G
101887-876EA 364.62 USD
101887-876 101887-878
3-(2-Chloro-5-methylphenoxy)azetidine
3-(2-Chloro-5-methylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
MDL Number: MFCD13559934
CAS Number: 1220028-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR