3-(2,5-Dichlorophenoxy)azetidine
Supplier: Matrix Scientific
049492-500MG
049492-2.5G
101887-872EA
364.62
USD
101887-872
101887-874
3-(2,5-Dichlorophenoxy)azetidine
3-(2,5-Dichlorophenoxy)azetidine
Hazard: Irritant
Formula:
C₉H₉Cl₂NO MW: 218.08 g/mol |
MDL Number:
MFCD12828795 CAS Number: 1236862-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...