3-(2,4,5-Trichlorophenoxy)azetidine

Supplier: Matrix Scientific

049491-500MG 049491-2.5G
101887-868EA 364.62 USD
101887-868 101887-870
3-(2,4,5-Trichlorophenoxy)azetidine
3-(2,4,5-Trichlorophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₈Cl₃NO
MDL Number: MFCD13559933
CAS Number: 1219982-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR