3-(2-Chloro-4-fluorophenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049489-500MG 049489-2.5G
101887-860EA 364.62 USD
101887-860 101887-862
3-(2-Chloro-4-fluorophenoxy)azetidine hydrochloride
3-(2-Chloro-4-fluorophenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₀Cl₂FNO
MDL Number: MFCD11849027
CAS Number: 900512-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR