3-(4-Bromo-2-chlorophenoxy)azetidine
Supplier: Matrix Scientific
049488-500MG
049488-2.5G
101887-856EA
364.62
USD
101887-856
101887-858
3-(4-Bromo-2-chlorophenoxy)azetidine
3-(4-Bromo-2-chlorophenoxy)azetidine
Hazard: Irritant
Formula:
C₉H₉BrClNO MW: 262.54 g/mol |
MDL Number:
MFCD09861974 CAS Number: 954226-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...