3-(2-Chlorophenoxy)azetidine

Supplier: Matrix Scientific

049486-500MG 049486-2.5G
101887-848EA 170.77 USD
101887-848 101887-850
3-(2-Chlorophenoxy)azetidine
3-(2-Chlorophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₁₀ClNO
MDL Number: MFCD09861863
CAS Number: 954222-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR