3-(2-Fluorophenoxy)azetidine

Supplier: Matrix Scientific

049485-500MG 049485-2.5G
101887-844EA 364.62 USD
101887-844 101887-846
3-(2-Fluorophenoxy)azetidine
3-(2-Fluorophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₁₀FNO
MDL Number: MFCD09861860
CAS Number: 918831-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR