3-(2-Bromo-4-methylphenoxy)azetidine

Supplier: Matrix Scientific

049484-500MG 049484-2.5G
101887-840EA 364.62 USD
101887-840 101887-842
3-(2-Bromo-4-methylphenoxy)azetidine
3-(2-Bromo-4-methylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD13559931
CAS Number: 1219982-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR