3-(2-Bromophenoxy)azetidine

Supplier: Matrix Scientific

049482-500MG 049482-2.5G
101887-832EA 364.62 USD
101887-832 101887-834
3-(2-Bromophenoxy)azetidine
3-(2-Bromophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₁₀BrNO
MW: 228.1 g/mol
MDL Number: MFCD09861869
CAS Number: 954223-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR