3-Azetidinyl phenyl ether hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-Phenoxyazetidine hydrochloride

049481-500MG 049481-2.5G
101887-828EA 124.62 USD
101887-828 101887-830
3-Azetidinyl phenyl ether hydrochloride
3-Azetidinyl phenyl ether hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClNO
MW: 185.66 g/mol
MDL Number: MFCD01314198
CAS Number: 301335-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR