3-(3-Methoxypropoxy)azetidine trifluoroacetate

Supplier: Matrix Scientific

049479-500MG 049479-2.5G
101887-820EA 364.62 USD
101887-820 101887-822
3-(3-Methoxypropoxy)azetidine trifluoroacetate
3-(3-Methoxypropoxy)azetidine trifluoroacetate

Hazard: Irritant

Formula: C₉H₁₆F₃NO₄
MDL Number: MFCD28291764
CAS Number: 1820736-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR