3-[(4-Methylbenzyl)oxy]azetidine

Supplier: Matrix Scientific

049475-500MG 049475-2.5G
101887-804EA 592.31 USD
101887-804 101887-806
3-[(4-Methylbenzyl)oxy]azetidine
3-[(4-Methylbenzyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD11874612
CAS Number: 1121633-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR