3-[(3-Methylbenzyl)oxy]azetidine

Supplier: Matrix Scientific

049469-500MG 049469-2.5G
101887-780EA 364.62 USD
101887-780 101887-782
3-[(3-Methylbenzyl)oxy]azetidine
3-[(3-Methylbenzyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD11874216
CAS Number: 1121628-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR