3-[(3-Chlorobenzyl)oxy]azetidine

Supplier: Matrix Scientific

049468-500MG 049468-2.5G
101887-776EA 364.62 USD
101887-776 101887-778
3-[(3-Chlorobenzyl)oxy]azetidine
3-[(3-Chlorobenzyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
MDL Number: MFCD11874739
CAS Number: 1121634-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR