3-[(3-Fluorobenzyl)oxy]azetidine

Supplier: Matrix Scientific

049467-500MG 049467-2.5G
101887-772EA 364.62 USD
101887-772 101887-774
3-[(3-Fluorobenzyl)oxy]azetidine
3-[(3-Fluorobenzyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₀H₁₂FNO
MDL Number: MFCD11874727
CAS Number: 1121634-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR