3-[(2,4-Dichlorobenzyl)oxy]azetidine

Supplier: Matrix Scientific

049464-500MG 049464-2.5G
101887-760EA 364.62 USD
101887-760 101887-762
3-[(2,4-Dichlorobenzyl)oxy]azetidine
3-[(2,4-Dichlorobenzyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₀H₁₁Cl₂NO
MDL Number: MFCD13559922
CAS Number: 1220038-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR