3-[(2-Chlorobenzyl)oxy]azetidine

Supplier: Matrix Scientific

Synonyms: 3-((2-Chlorobenzyl)oxy)azetidine

049463-500MG 049463-2.5G
101887-756EA 364.62 USD
101887-756 101887-758
3-[(2-Chlorobenzyl)oxy]azetidine
3-[(2-Chlorobenzyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
MW: 197.67 g/mol
MDL Number: MFCD11874528
CAS Number: 1121627-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR