3-(Benzyloxy)azetidine trifluoroacetate

Supplier: Matrix Scientific

049461-500MG 049461-2.5G
101887-748EA 384.62 USD
101887-748 101887-750
3-(Benzyloxy)azetidine trifluoroacetate
3-(Benzyloxy)azetidine trifluoroacetate

Hazard: Irritant

Formula: C₁₂H₁₄F₃NO₃
MDL Number: MFCD21605755
CAS Number: 1361115-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR