3-Azetidinyl ethyl ether trifluoroacetate

Supplier: Matrix Scientific

049459-500MG 049459-2.5G
101887-740EA 364.62 USD
101887-740 101887-742
3-Azetidinyl ethyl ether trifluoroacetate
3-Azetidinyl ethyl ether trifluoroacetate

Hazard: Irritant

Formula: C₇H₁₂F₃NO₃
MDL Number: MFCD29477422
CAS Number: 88536-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR