3-(3-Phenylpropoxy)azetidine

Supplier: Matrix Scientific

049455-500MG 049455-2.5G
101887-724EA 364.62 USD
101887-724 101887-726
3-(3-Phenylpropoxy)azetidine
3-(3-Phenylpropoxy)azetidine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MW: 191.28 g/mol
MDL Number: MFCD13559919
CAS Number: 1219982-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR