3-Azetidinyl propyl ether

Supplier: Matrix Scientific

049454-500MG 049454-2.5G
101887-720EA 364.62 USD
101887-720 101887-722
3-Azetidinyl propyl ether
3-Azetidinyl propyl ether

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD09701258
CAS Number: 897086-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR