3-(Isopentyloxy)azetidinetrifluoroacetic acid salt ≥95%

Supplier: Matrix Scientific

049453-500MG 049453-2.5G
101887-716EA 364.62 USD
101887-716 101887-718
3-(Isopentyloxy)azetidinetrifluoroacetic acid salt ≥95%
3-(Isopentyloxy)azetidinetrifluoroacetic acid salt

Hazard: Irritant

Formula: C₁₀H₁₈F₃NO₃
MDL Number: MFCD22373632
CAS Number: 1380571-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR