3-(Allyloxy)azetidine

Supplier: Matrix Scientific

049452-500MG 049452-2.5G
101887-712EA 384.62 USD
101887-712 101887-714
3-(Allyloxy)azetidine
3-(Allyloxy)azetidine

Hazard: Irritant

Formula: C₆H₁₁NO
MDL Number: MFCD11848752
CAS Number: 1489636-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR