3-(2-Propynyloxy)azetidine trifluoroacetate

Supplier: Matrix Scientific

049451-500MG 049451-2.5G
101887-708EA 364.62 USD
101887-708 101887-710
3-(2-Propynyloxy)azetidine trifluoroacetate
3-(2-Propynyloxy)azetidine trifluoroacetate

Hazard: Irritant

Formula: C₈H₁₀F₃NO₃
MDL Number: MFCD28976345
CAS Number: 1864051-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR