3-Azetidinyl-2-(2-methoxyethoxy)ethyl ether trifluoroacetate

Supplier: Matrix Scientific

049450-500MG 049450-2.5G
101887-704EA 364.62 USD
101887-704 101887-706
3-Azetidinyl-2-(2-methoxyethoxy)ethyl ether trifluoroacetate
3-Azetidinyl-2-(2-methoxyethoxy)ethyl ether trifluoroacetate

Hazard: Irritant

Formula: C₁₀H₁₈F₃NO₅
MDL Number: MFCD29925633
CAS Number: 1220038-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR