3-(2-Fluoroethoxy)azetidine trifluoroacetate salt

Supplier: Matrix Scientific

049446-500MG 049446-2.5G
101887-688EA 364.62 USD
101887-688 101887-690
3-(2-Fluoroethoxy)azetidine trifluoroacetate salt
3-(2-Fluoroethoxy)azetidine trifluoroacetate salt

Hazard: Irritant

Formula: C₇H₁₁F₄NO₃
MDL Number: MFCD22415176
CAS Number: 1401425-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR