3-(2-Cyclohexylethoxy)azetidine

Supplier: Matrix Scientific

049445-500MG 049445-2.5G
101887-684EA 364.62 USD
101887-684 101887-686
3-(2-Cyclohexylethoxy)azetidine
3-(2-Cyclohexylethoxy)azetidine

Hazard: Irritant

Formula: C₁₁H₂₁NO
MW: 183.3 g/mol
MDL Number: MFCD13559912
CAS Number: 1220038-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR