3-(Cyclopropylmethoxy)azetidine trifluoroacetate

Supplier: Matrix Scientific

049444-500MG 049444-2.5G
101887-680EA 384.62 USD
101887-680 101887-682
3-(Cyclopropylmethoxy)azetidine trifluoroacetate
3-(Cyclopropylmethoxy)azetidine trifluoroacetate

Hazard: Irritant

Formula: C₉H₁₄F₃NO₃
MDL Number: MFCD22378811
CAS Number: 1393330-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR