1-[4-(3-Azetidinyloxy)phenyl]-1-ethanone

Supplier: Matrix Scientific

049442-500MG 049442-2.5G
101887-672EA 364.62 USD
101887-672 101887-674
1-[4-(3-Azetidinyloxy)phenyl]-1-ethanone
1-[4-(3-Azetidinyloxy)phenyl]-1-ethanone

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD13559910
CAS Number: 1220021-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR