1-[2-(3-Azetidinyloxy)phenyl]-1-ethanone

Supplier: Matrix Scientific

049441-500MG 049441-2.5G
101887-668EA 364.62 USD
101887-668 101887-670
1-[2-(3-Azetidinyloxy)phenyl]-1-ethanone
1-[2-(3-Azetidinyloxy)phenyl]-1-ethanone

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD13559909
CAS Number: 1219948-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR