8-(3-Azetidinyloxy)-5-chloroquinoline
Supplier: Matrix Scientific
049440-500MG
049440-2.5G
101887-664EA
364.62
USD
101887-664
101887-666
8-(3-Azetidinyloxy)-5-chloroquinoline
8-(3-Azetidinyloxy)-5-chloroquinoline
Hazard: Irritant
Formula:
C₁₂H₁₁ClN₂O |
MDL Number:
MFCD13559908 CAS Number: 1219981-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...