3-(4-Bromo-2-nitrophenoxy)azetidine

Supplier: Matrix Scientific

049439-500MG 049439-2.5G
101887-660EA 364.62 USD
101887-660 101887-662
3-(4-Bromo-2-nitrophenoxy)azetidine
3-(4-Bromo-2-nitrophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₉BrN₂O₃
MDL Number: MFCD13559907
CAS Number: 1220027-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR