3-(4-Chloro-2-nitrophenoxy)azetidine

Supplier: Matrix Scientific

049438-500MG 049438-2.5G
101887-656EA 364.62 USD
101887-656 101887-658
3-(4-Chloro-2-nitrophenoxy)azetidine
3-(4-Chloro-2-nitrophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₉ClN₂O₃
MDL Number: MFCD13559906
CAS Number: 1219948-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR