Ethyl-4-(3-azetidinyloxy)benzoate

Supplier: Matrix Scientific

049437-500MG 049437-2.5G
101887-652EA 364.62 USD
101887-652 101887-654
Ethyl-4-(3-azetidinyloxy)benzoate
Ethyl-4-(3-azetidinyloxy)benzoate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD09861929
CAS Number: 954224-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR