Methyl-2-(3-azetidinyloxy)benzoate

Supplier: Matrix Scientific

049436-500MG 049436-2.5G
101887-648EA 364.62 USD
101887-648 101887-650
Methyl-2-(3-azetidinyloxy)benzoate
Methyl-2-(3-azetidinyloxy)benzoate

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD13559905
CAS Number: 1220038-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR