3-(3-Azetidinyloxy)-N,N-diethylaniline

Supplier: Matrix Scientific

049435-500MG 049435-2.5G
101887-644EA 364.62 USD
101887-644 101887-646
3-(3-Azetidinyloxy)-N,N-diethylaniline
3-(3-Azetidinyloxy)-N,N-diethylaniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂O
MDL Number: MFCD13559904
CAS Number: 1219960-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR