3-(4-Allyl-2-methoxyphenoxy)azetidine

Supplier: Matrix Scientific

049433-500MG 049433-2.5G
101887-636EA 364.62 USD
101887-636 101887-638
3-(4-Allyl-2-methoxyphenoxy)azetidine
3-(4-Allyl-2-methoxyphenoxy)azetidine

Hazard: Irritant

Formula: C₁₃H₁₇NO₂
MDL Number: MFCD13559902
CAS Number: 1219960-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR