3-Azetidinyl-4-(2-methoxyethyl)phenyl ether

Supplier: Matrix Scientific

049432-500MG 049432-2.5G
101887-632EA 364.62 USD
101887-632 101887-634
3-Azetidinyl-4-(2-methoxyethyl)phenyl ether
3-Azetidinyl-4-(2-methoxyethyl)phenyl ether

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD13559901
CAS Number: 1220027-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR